Ethyl-1H-indazole-3-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 1H-indazole-3-carboxylate

AS21160G1 AS21160G25 AS21160G5
ACTIAS21160G1EA 56 GBP
ACTIAS21160G1 ACTIAS21160G25 ACTIAS21160G5
Ethyl-1H-indazole-3-carboxylate
Ethyl 1H-indazole-3-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD01138134
CAS Number: 4498-68-4
EINECS: 224-795-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR