3-Amino-5-bromo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-1H-indazol-3-amine

AS22564G1 AS22564G25 AS22564G5
ACTIAS22564G1EA 16 GBP
ACTIAS22564G1 ACTIAS22564G25 ACTIAS22564G5
3-Amino-5-bromo-1H-indazole
3-Amino-5-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD03426696
CAS Number: 61272-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR