7-Bromo-3-iodo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12287G1 AS12287G5
ACTIAS12287G1EA 68 GBP
ACTIAS12287G1 ACTIAS12287G5
7-Bromo-3-iodo-1H-indazole
7-Bromo-3-iodo-1H-indazole
Formula: C₇H₄BrIN₂
MW: 322.9285 g/mol
MDL Number: MFCD08689979
CAS Number: 944904-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR