6-Bromo-3-iodo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12286G1 AS12286G5
ACTIAS12286G1EA 18 GBP
ACTIAS12286G1 ACTIAS12286G5
6-Bromo-3-iodo-1H-indazole
6-Bromo-3-iodo-1H-indazole
Formula: C₇H₄BrIN₂
MW: 322.9285 g/mol
MDL Number: MFCD07781599
CAS Number: 885521-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR