4-Bromo-6-fluoro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21178G1 AS21178G5
ACTIAS21178G1EA 44 GBP
ACTIAS21178G1 ACTIAS21178G5
4-Bromo-6-fluoro-1H-indazole
4-Bromo-6-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
MDL Number: MFCD07781448
CAS Number: 885520-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR