Methyl-6-bromo-1H-indazole-4-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24648G1 AS24648G5
ACTIAS24648G1EA 0 GBP
ACTIAS24648G1 ACTIAS24648G5
Methyl-6-bromo-1H-indazole-4-carboxylate
Methyl 6-bromo-1H-indazole-4-carboxylate
Formula: C₉H₇BrN₂O₂
MDL Number: MFCD07781319
CAS Number: 885518-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR