Methyl-3-bromo-1H-indazole-6-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12254G1 AS12254G5
ACTIAS12254G1EA 0 GBP
ACTIAS12254G1 ACTIAS12254G5
Methyl-3-bromo-1H-indazole-6-carboxylate
Methyl 3-bromo-1H-indazole-6-carboxylate
Formula: C₉H₇BrN₂O₂
MDL Number: MFCD11499017
CAS Number: 192945-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR