4-Bromo-6-chloro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21177G1 AS21177G5
ACTIAS21177G1EA 0 GBP
ACTIAS21177G1 ACTIAS21177G5
4-Bromo-6-chloro-1H-indazole
4-Bromo-6-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
MDL Number: MFCD07781352
CAS Number: 885519-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR