1H-Indazole-6-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22378G1 AS22378G5
ACTIAS22378G1EA 0 GBP
ACTIAS22378G1 ACTIAS22378G5
1H-Indazole-6-boronic acid pinacol ester
1H-Indazole-6-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD07368067
CAS Number: 937049-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR