1H-Indazole-4-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21215G1 AS21215G5
ACTIAS21215G1EA 0 GBP
ACTIAS21215G1 ACTIAS21215G5
1H-Indazole-4-boronic acid pinacol ester
1H-Indazole-4-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD08059267
CAS Number: 885618-33-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR