5-Bromobenzoxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromobenzo[d]oxazole

HI-1573-5G HI-1573-1G
COBBHI-1573-5GEA 0 GBP
COBBHI-1573-5G COBBHI-1573-1G
5-Bromobenzoxazole
5-Bromobenzoxazole
Formula: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD03095026
CAS Number: 132244-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR