3-Chloro-6-(propylamino)pyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6091-5G HC-6091-1G
COBBHC-6091-5GEA 0 GBP
COBBHC-6091-5G COBBHC-6091-1G
3-Chloro-6-(propylamino)pyridazine
3-Chloro-6-(propylamino)pyridazine
Formula: C₇H₁₀ClN₃
MDL Number: MFCD09800940
CAS Number: 951885-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR