3-(Butylamino)-6-chloropyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6142-5G HC-6142-1G
COBBHC-6142-5GEA 0 GBP
COBBHC-6142-5G COBBHC-6142-1G
3-(Butylamino)-6-chloropyridazine
3-(Butylamino)-6-chloropyridazine
Formula: C₈H₁₂ClN₃
MDL Number: MFCD09800957
CAS Number: 1009-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR