3-Amino-4-bromo-6-chloropyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-6-chloro-pyridazin-3-amine

HC-6742-5G HC-6742-1G
COBBHC-6742-5GEA 0 GBP
COBBHC-6742-5G COBBHC-6742-1G
3-Amino-4-bromo-6-chloropyridazine
3-Amino-4-bromo-6-chloropyridazine
Formula: C₄H₃BrClN₃
MW: 208.4437 g/mol
MDL Number: MFCD11520891
CAS Number: 446273-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR