Ethyl-5-amino-4-cyano-1-(2-methylphenyl)-1H-pyrazole-3-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2532-1G HC-2532-250M
COBBHC-2532-1GEA 0 GBP
COBBHC-2532-1G COBBHC-2532-250M
Ethyl-5-amino-4-cyano-1-(2-methylphenyl)-1H-pyrazole-3-carboxylate
Ethyl 5-amino-4-cyano-1-(2-methylphenyl)-1H-pyrazole-3-carboxylate
Formula: C₁₄H₁₄N₄O₂
MDL Number: MFCD09835733
CAS Number: 1150164-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR