3-(3-Chlorophenyl)-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6892-1G HC-6892-5G
COBBHC-6892-1GEA 0 GBP
COBBHC-6892-1G COBBHC-6892-5G
3-(3-Chlorophenyl)-1H-pyrazole
3-(3-Chlorophenyl)-1H-pyrazole
Formula: C₉H₇ClN₂
MDL Number: MFCD09860599
CAS Number: 59843-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR