4-Bromo-1-(2-nitrophenyl)-1H-pyrazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1314-5G
HI-1314-1G
COBBHI-1314-5GEA
0
GBP
COBBHI-1314-5G
COBBHI-1314-1G
4-Bromo-1-(2-nitrophenyl)-1H-pyrazole
4-Bromo-1-(2-nitrophenyl)-1H-pyrazole
Formula:
C₉H₆BrN₃O₂ |
MDL Number:
MFCD09475850 CAS Number: 957034-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...