4-Bromo-1-(2-fluorophenyl)-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1337-1G HI-1337-5G
COBBHI-1337-1GEA 0 GBP
COBBHI-1337-1G COBBHI-1337-5G
4-Bromo-1-(2-fluorophenyl)-1H-pyrazole
4-Bromo-1-(2-fluorophenyl)-1H-pyrazole
Formula: C₉H₆BrFN₂
MDL Number: MFCD09878403
CAS Number: 957062-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR