4-Bromo-1-phenyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1704-5G HI-1704-1G
COBBHI-1704-5GEA 0 GBP
COBBHI-1704-5G COBBHI-1704-1G
4-Bromo-1-phenyl-1H-pyrazole
4-Bromo-1-phenyl-1H-pyrazole
Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD00234505
CAS Number: 15115-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR