5-Methyl-4,5,6,7-tetrahydro-2H-indazole hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3316-5G HC-3316-1G
COBBHC-3316-5GEA 0 GBP
COBBHC-3316-5G COBBHC-3316-1G
5-Methyl-4,5,6,7-tetrahydro-2H-indazole hydrochloride
5-Methyl-4,5,6,7-tetrahydro-2H-indazole hydrochloride
Formula: C₈H₁₃ClN₂
MDL Number: MFCD16295092
CAS Number: 1226808-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR