4,5,6,7-Tetrahydro-1H-indazole hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AM-1740-25G
AM-1740-5G
AM-1740-1G
COBBAM-1740-25GEA
0
GBP
COBBAM-1740-25G
COBBAM-1740-5G
COBBAM-1740-1G
4,5,6,7-Tetrahydro-1H-indazole hydrochloride
4,5,6,7-Tetrahydro-1H-indazole hydrochloride
Formula:
C₇H₁₁ClN₂ |
MDL Number:
MFCD13195636 CAS Number: 18161-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...