5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11F-419S.500
11F-419S.1
11F-419S.5
11F-419S.10
BIOT11F-419S.500EA
0
GBP
BIOT11F-419S.500
BIOT11F-419S.1
BIOT11F-419S.5
BIOT11F-419S.10
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Formula:
C₉H₁₁ClN₂O MW: 158.59 g/mol |
MDL Number:
MFCD00052538 CAS Number: 27006-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...