Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate, Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate

005979-1G 005979-5G 5979
FLUO005979-1GEA 0 GBP
FLUO005979-1G FLUO005979-5G FLUO5979
Ethyl-5-amino-3-methyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate
Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD01210846
CAS Number: 23286-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR