Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21355G100 AS21355G25
ACTIAS21355G100EA 0 GBP
ACTIAS21355G100 ACTIAS21355G25
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Formula: C₇H₁₁N₃O₂
MDL Number: MFCD00051652
CAS Number: 31037-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR