1-Boc-7-methylindole-3-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-6319-1G PN-6319-250M
COBBPN-6319-1GEA 0 GBP
COBBPN-6319-1G COBBPN-6319-250M
1-Boc-7-methylindole-3-boronic acid pinacol ester
1-Boc-7-methylindole-3-boronic acid pinacol ester
Formula: C₂₀H₂₈BNO₄
MDL Number: MFCD16295117
CAS Number: 1256360-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR