1-Boc-6-chloroindole-3-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl 6-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

PN-6217-1G PN-6217-250M
COBBPN-6217-1GEA 0 GBP
COBBPN-6217-1G COBBPN-6217-250M
1-Boc-6-chloroindole-3-boronic acid pinacol ester
1-Boc-6-chloroindole-3-boronic acid pinacol ester
Formula: C₁₉H₂₅BClNO₄
MW: 377.6701 g/mol
MDL Number: MFCD15143606
CAS Number: 1218790-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR