1-Boc-5-(methoxycarbonyl)indole-2-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5316-5G BB-5316-1G BB-5316-250M
COBBBB-5316-5GEA 0 GBP
COBBBB-5316-5G COBBBB-5316-1G COBBBB-5316-250M
1-Boc-5-(methoxycarbonyl)indole-2-boronic acid
1-Boc-5-(methoxycarbonyl)indole-2-boronic acid
Formula: C₁₅H₁₈BNO₆
MDL Number: MFCD12756415
CAS Number: 1217500-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR