1-Boc-4-(ethoxycarbonyl)indole-2-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5430-1G BB-5430-250M
COBBBB-5430-1GEA 221.59 GBP
COBBBB-5430-1G COBBBB-5430-250M
1-Boc-4-(ethoxycarbonyl)indole-2-boronic acid
1-Boc-4-(ethoxycarbonyl)indole-2-boronic acid
Formula: C₁₆H₂₀BNO₆
MDL Number: MFCD11855837
CAS Number: 1150114-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR