5-Cyanoindole-2-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2364-1G BB-2364-250M
COBBBB-2364-1GEA 0 GBP
COBBBB-2364-1G COBBBB-2364-250M
5-Cyanoindole-2-boronic acid
5-Cyanoindole-2-boronic acid
Formula: C₉H₇BN₂O₂
MW: 185.98 g/mol
MDL Number: MFCD06798223
CAS Number: 871329-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR