6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7164-5G PY-7164-1G
COBBPY-7164-5GEA 0 GBP
COBBPY-7164-5G COBBPY-7164-1G
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one
Formula: C₉H₃Cl₂F₃N₂O
MDL Number: MFCD09801051
CAS Number: 477857-25-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR