6,7-Dichloro-3-(trifluoromethyl)-2-quinoxalinecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CS-9161-250M CS-9161-100M
COBBCS-9161-250MEA 0 GBP
COBBCS-9161-250M COBBCS-9161-100M
6,7-Dichloro-3-(trifluoromethyl)-2-quinoxalinecarboxylic acid
6,7-Dichloro-3-(trifluoromethyl)-2-quinoxalinecarboxylic acid
Formula: C₁₀H₃Cl₂F₃N₂O₂
MDL Number: MFCD09800927
CAS Number: 951884-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR