6-(Trifluoromethoxy)quinoxaline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-3081-5G
HC-3081-1G
COBBHC-3081-5GEA
0
GBP
COBBHC-3081-5G
COBBHC-3081-1G
6-(Trifluoromethoxy)quinoxaline hydrochloride
6-(Trifluoromethoxy)quinoxaline hydrochloride
Formula:
C₉H₆ClF₃N₂O |
MDL Number:
MFCD13195695 CAS Number: 1215206-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...