6-(Trifluoromethoxy)quinoxaline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1304-5G PY-1304-1G
COBBPY-1304-5GEA 0 GBP
COBBPY-1304-5G COBBPY-1304-1G
6-(Trifluoromethoxy)quinoxaline
6-(Trifluoromethoxy)quinoxaline
Formula: C₉H₅F₃N₂O
MDL Number: MFCD13180522
CAS Number: 1215205-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR