2-Quinoxalinecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20333G100 AS20333G25 AS20333G5
ACTIAS20333G100EA 191 GBP
ACTIAS20333G100 ACTIAS20333G25 ACTIAS20333G5
2-Quinoxalinecarboxylic acid
2-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
MDL Number: MFCD00012334
CAS Number: 879-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR