Methyl quinoxaline-6-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 6-Quinoxalinecarboxylate

AS22095G25 AS22095G5
ACTIAS22095G25EA 299 GBP
ACTIAS22095G25 ACTIAS22095G5
Methyl quinoxaline-6-carboxylate
Methyl quinoxaline-6-carboxylate
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00102618
CAS Number: 23088-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR