6-Quinoxaline​boronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

708631-250MG 708631-1G
SIAL708631-250MGEA 96.6 GBP
SIAL708631-250MG SIAL708631-1G
6-Quinoxaline​boronic acid pinacol ester, Sigma-Aldrich®
6-Quinoxaline​boronic acid pinacol ester
Formula: C₁₄H₁₇BN₂O₂
MW: 256.1080 g/mol
MDL Number: MFCD11054040
CAS Number: 1167418-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR