6-Fluoro-1-phenylbenzimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0200-1G IN-0200-5G
COBBIN-0200-1GEA 0 GBP
COBBIN-0200-1G COBBIN-0200-5G
6-Fluoro-1-phenylbenzimidazole
6-Fluoro-1-phenylbenzimidazole
Formula: C₈H₇FN₂
MDL Number: MFCD12546505
CAS Number: 1187385-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR