6-Methyl-3-isoquinolinol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methyl-3-hydroxyisoquinoline

HC-6668-1G HC-6668-5G
COBBHC-6668-1GEA 0 GBP
COBBHC-6668-1G COBBHC-6668-5G
6-Methyl-3-isoquinolinol
6-Methyl-3-isoquinolinol
Formula: C₁₀H₉NO
MDL Number: MFCD12546459
CAS Number: 51463-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR