1-Methyl-3-isoquinolinol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methylisoquinolin-3-ol

HC-6874-1G HC-6874-5G
COBBHC-6874-1GEA 0 GBP
COBBHC-6874-1G COBBHC-6874-5G
1-Methyl-3-isoquinolinol
1-Methyl-3-isoquinolinol
Formula: C₁₀H₉NO
MDL Number: MFCD11053887
CAS Number: 16535-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR