3-Chloro-6-(pyrrol-1-yl)pyridazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6099-5G
HC-6099-1G
COBBHC-6099-5GEA
0
GBP
COBBHC-6099-5G
COBBHC-6099-1G
3-Chloro-6-(pyrrol-1-yl)pyridazine
3-Chloro-6-(pyrrol-1-yl)pyridazine
Formula:
C₈H₆ClN₃ |
MDL Number:
MFCD00828665 CAS Number: 5096-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...