3-Chloro-6-(1H-pyrazol-1-yl)pyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6094-5G HC-6094-1G
COBBHC-6094-5GEA 0 GBP
COBBHC-6094-5G COBBHC-6094-1G
3-Chloro-6-(1H-pyrazol-1-yl)pyridazine
3-Chloro-6-(1H-pyrazol-1-yl)pyridazine
Formula: C₇H₅ClN₄
MDL Number: MFCD09261072
CAS Number: 29334-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR