3-(4-Bromo-1H-pyrazol-1-yl)-6-chloropyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6095-5G HC-6095-1G
COBBHC-6095-5GEA 0 GBP
COBBHC-6095-5G COBBHC-6095-1G
3-(4-Bromo-1H-pyrazol-1-yl)-6-chloropyridazine
3-(4-Bromo-1H-pyrazol-1-yl)-6-chloropyridazine
Formula: C₇H₄BrClN₄
MDL Number: MFCD09475884
CAS Number: 957035-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR