1-(6-Chloro-3-pyridazinyl)piperidin-4-ol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22012G1 AS22012G25 AS22012G5
ACTIAS22012G1EA 74 GBP
ACTIAS22012G1 ACTIAS22012G25 ACTIAS22012G5
1-(6-Chloro-3-pyridazinyl)piperidin-4-ol
1-(6-Chloro-3-pyridazinyl)piperidin-4-ol
Formula: C₉H₁₂ClN₃O
MDL Number: MFCD05864736
CAS Number: 89937-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR