3-Chloro-6-(pyrrolidin-1-yl)pyridazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6045-5G
HC-6045-1G
HC-6045-25G
COBBHC-6045-5GEA
0
GBP
COBBHC-6045-5G
COBBHC-6045-1G
COBBHC-6045-25G
3-Chloro-6-(pyrrolidin-1-yl)pyridazine
3-Chloro-6-(pyrrolidin-1-yl)pyridazine
Formula:
C₈H₁₀ClN₃ |
MDL Number:
MFCD05864735 CAS Number: 66346-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...