3-(Benzyloxy)-6-methylpyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1395-1G PY-1395-5G
COBBPY-1395-1GEA 0 GBP
COBBPY-1395-1G COBBPY-1395-5G
3-(Benzyloxy)-6-methylpyridazine
3-(Benzyloxy)-6-methylpyridazine
Formula: C₁₂H₁₂N₂O
MDL Number: MFCD16036159
CAS Number: 6958-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR