Pyrazolo[1,5-a]pyridine-2-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21448G1 AS21448G5
ACTIAS21448G1EA 49 GBP
ACTIAS21448G1 ACTIAS21448G5
Pyrazolo[1,5-a]pyridine-2-carboxylic acid
Pyrazolo[1,5-a]pyridine-2-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1500 g/mol
MDL Number: MFCD06739057
CAS Number: 63237-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR