Methyl pyrazolo[1,5-a]pyridine-3-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pyrazolo[1,5-a]pyridine-3-carboxylic acid methyl ester

AS12054G1 AS12054G5
ACTIAS12054G1EA 155 GBP
ACTIAS12054G1 ACTIAS12054G5
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate
Methyl pyrazolo[1,5-a]pyridine-3-carboxylate
Formula: C₉H₈N₂O₂
MDL Number: MFCD09752651
CAS Number: 63237-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR