(3,5-Dimethyl-4-isoxazolyl)methanol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,5-Dimethyl-4-(hydroxymethyl)isoxazole
AS21531G1
AS21531G25
AS21531G5
ACTIAS21531G1EA
84
GBP
ACTIAS21531G1
ACTIAS21531G25
ACTIAS21531G5
(3,5-Dimethyl-4-isoxazolyl)methanol
(3,5-Dimethyl-4-isoxazolyl)methanol
Formula:
C₆H₉NO₂ MW: 127.1400 g/mol |
MDL Number:
MFCD00464226 CAS Number: 19788-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...