1-Methyl-6-nitro-1H-indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS12529G1
AS12529G5
ACTIAS12529G1EA
37
GBP
ACTIAS12529G1
ACTIAS12529G5
1-Methyl-6-nitro-1H-indazole
1-Methyl-6-nitro-1H-indazole
Formula:
C₈H₇N₃O₂ MW: 177.160 g/mol |
MDL Number:
MFCD00456124 CAS Number: 6850-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...