6-(Trifluoromethyl)-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6972-5G
HC-6972-1G
COBBHC-6972-5GEA
0
GBP
COBBHC-6972-5G
COBBHC-6972-1G
6-(Trifluoromethyl)-1H-indazole
6-(Trifluoromethyl)-1H-indazole
Formula:
C₈H₅F₃N₂ |
MDL Number:
MFCD08690097 CAS Number: 954239-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...